Learn More
Thiocarbohydrazide, 98%
CAS: 2231-57-4 | CH6N4S | 106.15 g/mol
$120.91 - $382.21
Chemical Identifiers
| CAS | 2231-57-4 |
|---|---|
| Molecular Formula | CH6N4S |
| Molecular Weight (g/mol) | 106.15 |
| MDL Number | MFCD00007616 |
| InChI Key | LJTFFORYSFGNCT-UHFFFAOYSA-N |
| Synonym | thiocarbohydrazide, thiocarbazide, thiocarbonohydrazide, carbonothioic dihydrazide, carbohydrazide, thio, thiocarbonic dihydrazide, 1,3-diamino-2-thiourea, hydrazinecarbohydrazonothioic acid, usaf ek-7372, unii-1iz2h82nwu |
| PubChem CID | 2724189 |
| ChEBI | CHEBI:9549 |
| IUPAC Name | 1,3-diaminothiourea |
| SMILES | NNC(=S)NN |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC207530050
|
Thermo Scientific Chemicals
207530050 |
5 g | Glass bottle |
Each for $120.91
|
|
||||
|
AC207530250
|
Thermo Scientific Chemicals
207530250 |
25 g | Plastic bottle |
Each for $382.21
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 2231-57-4 | |
| 106.15 | |
| LJTFFORYSFGNCT-UHFFFAOYSA-N | |
| 2724189 | |
| 1,3-diaminothiourea |
| CH6N4S | |
| MFCD00007616 | |
| thiocarbohydrazide, thiocarbazide, thiocarbonohydrazide, carbonothioic dihydrazide, carbohydrazide, thio, thiocarbonic dihydrazide, 1,3-diamino-2-thiourea, hydrazinecarbohydrazonothioic acid, usaf ek-7372, unii-1iz2h82nwu | |
| CHEBI:9549 | |
| NNC(=S)NN |
Specifications
| 2231-57-4 | |
| Beige-Gray to White | |
| 97.5% min | |
| CH6N4S | |
| MFCD00007616 | |
| 03, 197 | |
| LJTFFORYSFGNCT-UHFFFAOYSA-N | |
| 1,3-diaminothiourea | |
| 2724189 | |
| 106.15 | |
| Crystalline Powder |
| 168.0°C to 172.0°C | |
| Authentic | |
| Glass bottle | |
| H2NNHCSNHNH2 | |
| 5 g | |
| thiocarbohydrazide, thiocarbazide, thiocarbonohydrazide, carbonothioic dihydrazide, carbohydrazide, thio, thiocarbonic dihydrazide, 1,3-diamino-2-thiourea, hydrazinecarbohydrazonothioic acid, usaf ek-7372, unii-1iz2h82nwu | |
| NNC(=S)NN | |
| 106.15 | |
| CHEBI:9549 | |
| 98% | |
| Thiocarbohydrazide, 98% |
Safety and Handling
GHS H Statement
Fatal if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Do not eat,drink or smoke when using th
GHS Signal Word: Danger
EINECSNumber : 218-769-8
RTECSNumber : FF2975000
TSCA : TSCA
RUO – Research Use Only