missing translation for 'onlineSavingsMsg'
Learn More

Thiocarbohydrazide, 97%

Catalog No. AAL0120506
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0120506 5 g
AAL0120514 25 g
AAL0120522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0120506 Supplier Thermo Scientific Chemicals Supplier No. L0120506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2231-57-4 | CH6N4S | 106.15 g/mol

Thiocarbohydrazide is used in electron microscopy to produce electron-opaque deposits for ultra structural analysis. It is used in the synthesis of oxazine grass ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiocarbohydrazide is used in electron microscopy to produce electron-opaque deposits for ultra structural analysis. It is used in the synthesis of oxazine grass ketones.

Solubility
Very slightly soluble (0.5g/100g, 25°C). Insoluble in ether.

Notes
Light Sensitive. Incompatible with strong acids, Strong oxidizing agents. Avoid heat.

Chemical Identifiers

CAS 2231-57-4
Molecular Formula CH6N4S
Molecular Weight (g/mol) 106.15
MDL Number MFCD00007616
InChI Key LJTFFORYSFGNCT-UHFFFAOYSA-N
Synonym thiocarbohydrazide, thiocarbazide, thiocarbonohydrazide, carbonothioic dihydrazide, carbohydrazide, thio, thiocarbonic dihydrazide, 1,3-diamino-2-thiourea, hydrazinecarbohydrazonothioic acid, usaf ek-7372, unii-1iz2h82nwu
PubChem CID 2724189
ChEBI CHEBI:9549
IUPAC Name 1,3-diaminothiourea
SMILES NNC(=S)NN

Specifications

Melting Point ∽175°C (decomposition)
Quantity 5 g
UN Number UN2811
Beilstein 506657
Sensitivity Light sensitive
Solubility Information Very slightly soluble (0.5g/100g,25°C). Insoluble in ether.
Formula Weight 106.15
Percent Purity 97%
Chemical Name or Material Thiocarbohydrazide
Hazard Category H300+H330-H311
Hazard Statement GHS H Statement
H300-H330
Fatal if swallowed.
Fatal if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P310-P312-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 218-769-8
RTECSNumber FF2975000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only