missing translation for 'onlineSavingsMsg'
Learn More

Thiobenzoic Acid 93.0+%, TCI America™

Catalog No. p-7136135
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T019525G 25 g
T0195500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T019525G Supplier TCI America Supplier No. T019525G
Only null left

Chemical Identifiers

CAS 98-91-9
Molecular Formula C7H6OS
Molecular Weight (g/mol) 138.184
MDL Number MFCD00004852
InChI Key UIJGNTRUPZPVNG-UHFFFAOYSA-N
Synonym thiobenzoic acid, benzenecarbothioic acid, benzoyl thiol, monothiobenzoic acid, benzoic acid, thio, acido mercaptobenzoico, unii-gbg5rlo56n, ccris 8913, acido mercaptobenzoico italian, gbg5rlo56n
PubChem CID 7414
IUPAC Name benzenecarbothioic S-acid
SMILES C1=CC=C(C=C1)C(=O)S

Specifications

Boiling Point 87°C
Quantity 25 g
Formula Weight 138.18
Percent Purity ≥93.0% (T)
Physical Form Liquid
Chemical Name or Material Thiobenzoic Acid
EINECSNumber (3)-1396
RTECSNumber DH6839000
TSCA Yes
Recommended Storage Refrigerator