missing translation for 'onlineSavingsMsg'
Learn More

Thiobenzamide 98.0+%, TCI America™

Catalog No. p-7136134
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T019425G 25 g
T0194100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T019425G Supplier TCI America Supplier No. T019425G
Only null left

Chemical Identifiers

CAS 2227-79-4
Molecular Formula C7H7NS
Molecular Weight (g/mol) 137.20
MDL Number MFCD00008060
InChI Key QIOZLISABUUKJY-UHFFFAOYSA-N
Synonym thiobenzamide, benzothioamide, benzamide, thio, benzothiamide, tiobenzamide, phenylthioamide, tiobenzamide italian, unii-8799vm9sxc, benzene-1-carbothioamide, aminophenylmethane-1-thione
PubChem CID 683563
ChEBI CHEBI:80418
IUPAC Name benzenecarbothioamide
SMILES NC(=S)C1=CC=CC=C1

Specifications

Melting Point 117°C
Color Yellow
Quantity 25 g
UN Number 2811
Formula Weight 137.20
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Thiobenzamide
RTECSNumber CV5860000
TSCA Yes