missing translation for 'onlineSavingsMsg'
Learn More

Thiobenzamide, 98%

CAS: 2227-79-4 | C7H7NS | 137.20 g/mol

Supplier:  Thermo Scientific Chemicals A1297336

 View more versions of this product

Catalog No. AAA1297336

Only null left
Explore available promotions

Description

Description

Thiobenzamide is useful for the preparation of amide and amidine adducts. It is also used to prepare 4-oxo-4H-chromene-3-carbothioic acid N-phenylamides. Further, it acts as a raw material in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiobenzamide is useful for the preparation of amide and amidine adducts. It is also used to prepare 4-oxo-4H-chromene-3-carbothioic acid N-phenylamides. Further, it acts as a raw material in organic synthesis.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications

Specifications

Thiobenzamide
114°C to 118°C
C7H7NS
500 g
606021
Slightly soluble in water.
NC(=S)C1=CC=CC=C1
137.20
CHEBI:80418
98%
2227-79-4
120°C (248°F)
MFCD00008060
UN2811
thiobenzamide, benzothioamide, benzamide, thio, benzothiamide, tiobenzamide, phenylthioamide, tiobenzamide italian, unii-8799vm9sxc, benzene-1-carbothioamide, aminophenylmethane-1-thione
QIOZLISABUUKJY-UHFFFAOYSA-N
benzenecarbothioamide
683563
137.21

Chemical Identifiers

2227-79-4
137.20
QIOZLISABUUKJY-UHFFFAOYSA-N
683563
benzenecarbothioamide
C7H7NS
MFCD00008060
thiobenzamide, benzothioamide, benzamide, thio, benzothiamide, tiobenzamide, phenylthioamide, tiobenzamide italian, unii-8799vm9sxc, benzene-1-carbothioamide, aminophenylmethane-1-thione
CHEBI:80418
NC(=S)C1=CC=CC=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H301-H315-H319-H332-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 218-765-6

RTECSNumber : CV5860000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only