missing translation for 'onlineSavingsMsg'
Learn More

Thiobenzamide, 95%

CAS: 2227-79-4 | C7H7NS | 137.20 g/mol

$52.73 - $52.73

Chemical Identifiers

CAS 2227-79-4
Molecular Formula C7H7NS
Molecular Weight (g/mol) 137.20
MDL Number MFCD00008060
InChI Key QIOZLISABUUKJY-UHFFFAOYSA-N
Synonym thiobenzamide, benzothioamide, benzamide, thio, benzothiamide, tiobenzamide, phenylthioamide, tiobenzamide italian, unii-8799vm9sxc, benzene-1-carbothioamide, aminophenylmethane-1-thione
PubChem CID 683563
ChEBI CHEBI:80418
IUPAC Name benzenecarbothioamide
SMILES NC(=S)C1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC164730100
SDS View Documents
Thermo Scientific Chemicals
164730100
10 g
Each for $52.73
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2227-79-4
137.20
QIOZLISABUUKJY-UHFFFAOYSA-N
683563
benzenecarbothioamide
C7H7NS
MFCD00008060
thiobenzamide, benzothioamide, benzamide, thio, benzothiamide, tiobenzamide, phenylthioamide, tiobenzamide italian, unii-8799vm9sxc, benzene-1-carbothioamide, aminophenylmethane-1-thione
CHEBI:80418
NC(=S)C1=CC=CC=C1

Specifications

2227-79-4
Yellow to Green
94% min. (HPLC)
C7H7NS
MFCD00008060
09, 424
Solubility in water: slightly soluble
NC(=S)C1=CC=CC=C1
137.20
CHEBI:80418
95%
Thiobenzamide, 95%
114°C to 118°C
Authentic
Glass bottle
C6H5CSNH2
10 g
thiobenzamide, benzothioamide, benzamide, thio, benzothiamide, tiobenzamide, phenylthioamide, tiobenzamide italian, unii-8799vm9sxc, benzene-1-carbothioamide, aminophenylmethane-1-thione
QIOZLISABUUKJY-UHFFFAOYSA-N
benzenecarbothioamide
683563
137.21
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Toxic if swallowed.
Causes skin irritation.
Harmful if inhaled.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 218-765-6

RTECSNumber : CV5860000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only