Learn More
Thioanisole, 99%
CAS: 100-68-5 | C7H8S | 124.201 g/mol
Supplier: Thermo Scientific Chemicals A1484622
Description
Thioanisole is used as an intermediate in the preparation of dyes, pharmaceuticals and agrochemicals. It is also used to prepare stable sulfonium salts, sulfoxides and sufones. It is utilized for the cleavage of methyl ethers in association with triflic acid. It acts as a flavoring agent or an adjuvant. It is an important starting material for the synthesis of 3-substituted benzo[b]thiophenes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsThioanisole is used as an intermediate in the preparation of dyes, pharmaceuticals and agrochemicals. It is also used to prepare stable sulfonium salts, sulfoxides and sufones. It is utilized for the cleavage of methyl ethers in association with triflic acid. It acts as a flavoring agent or an adjuvant. It is an important starting material for the synthesis of 3-substituted benzo[b]thiophenes.
Solubility
Miscible with most common organic solvents. Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
Specifications
| Thioanisole | |
| -15°C | |
| 188°C to 189°C | |
| Stench | |
| 1.586 | |
| 100 g | |
| 1904179 | |
| Miscible with most common organic solvents. Immiscible with water. | |
| CSC1=CC=CC=C1 | |
| 124.201 | |
| 124.21 |
| 100-68-5 | |
| 1.058 | |
| 73°C (163°F) | |
| C7H8S | |
| MFCD00008559 | |
| UN1993 | |
| thioanisole, methyl phenyl sulfide, methylthio benzene, thioanisol, benzene, methylthio, phenylthiomethane, phenyl methyl sulfide, methylsulfanyl benzene, methylphenylsulfide, 1-thiaethyl benzene | |
| HNKJADCVZUBCPG-UHFFFAOYSA-N | |
| methylsulfanylbenzene | |
| 7520 | |
| 99% |
Chemical Identifiers
| 100-68-5 | |
| 124.201 | |
| HNKJADCVZUBCPG-UHFFFAOYSA-N | |
| 7520 | |
| CSC1=CC=CC=C1 |
| C7H8S | |
| MFCD00008559 | |
| thioanisole, methyl phenyl sulfide, methylthio benzene, thioanisol, benzene, methylthio, phenylthiomethane, phenyl methyl sulfide, methylsulfanyl benzene, methylphenylsulfide, 1-thiaethyl benzene | |
| methylsulfanylbenzene |
Safety and Handling
GHS H Statement
H226-H302-H315-H319-H335
Flammable liquid and vapour.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P330-P333+P313-P362-P370+P378q-P501c
H227-H302-H315-H317-H319
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 202-878-2
RTECSNumber : DA6200000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only