missing translation for 'onlineSavingsMsg'
Learn More

Thioacetic Acid 95.0+%, TCI America™

Catalog No. T018925ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T018925ML 25 mL
T0189100ML 100 mL
T0189500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T018925ML Supplier TCI America Supplier No. T018925ML

Chemical Identifiers

CAS 507-09-5
Molecular Formula C2H4OS
Molecular Weight (g/mol) 76.11
MDL Number MFCD00004853
InChI Key DUYAAUVXQSMXQP-UHFFFAOYSA-N
Synonym thioacetic acid, ethanethioic acid, thiacetic acid, thioacetic s-acid, acetyl mercaptan, ethanethiolic acid, thiolacetic acid, thionoacetic acid, methanecarbothiolic acid, ch3cosh
PubChem CID 10484
ChEBI CHEBI:16555
IUPAC Name ethanethioic S-acid
SMILES CC(S)=O

Specifications

Color Yellow
Boiling Point 93°C
Quantity 25 mL
UN Number 2436
Formula Weight 76.11
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Thioacetic Acid
EINECSNumber (2)-0691
RTECSNumber AJ5600000
TSCA Yes
Recommended Storage Refrigerator