accessibility menu, dialog, popup

UserName

CAS RN 507-09-5

IUPAC Name: ethanethioic S-acid
Synonyms: schiff reagent potassium salt thioacetic acid thiacetic acid methanecarbothiolic acid kyselina thiooctova thionoacetic acid ethanethioic S-acid thioacetic S-acid schiff's reagent ethanethioic acid
Molecular Weight (g/mol): 76.11
Molecular Formula: C2H4OS
InChi Key: DUYAAUVXQSMXQP-UHFFFAOYSA-N
SMILES: CC(S)=O
Boiling Point 
  • (3)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (6)