Learn More
Thioacetic acid, 98%
CAS: 507-09-5 | C2H4OS | 76.11 g/mol
Supplier: Thermo Scientific Chemicals 138781000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Thioacetic acid | |
| 507-09-5 | |
| Colorless to Yellow | |
| 87.0°C to 93.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4630 to 1.4650 | |
| MFCD00004853 | |
| 02, 230 | |
| 1.07 | |
| thioacetic acid, ethanethioic acid, thiacetic acid, thioacetic s-acid, acetyl mercaptan, ethanethiolic acid, thiolacetic acid, thionoacetic acid, methanecarbothiolic acid, ch3cosh | |
| DUYAAUVXQSMXQP-UHFFFAOYSA-N | |
| ethanethioic S-acid | |
| 10484 | |
| 76.11 | |
| Liquid |
| 98% | |
| -17.0°C | |
| 1.0700g/mL | |
| 18°C | |
| 97.5% min. (Argentometry) | |
| C2H4OS | |
| CH3COSH | |
| 100 g | |
| 01,1154 | |
| 15, 9473 | |
| Solubility in water: 27g/L water (15°C) (hydrolyse). Other solubilities: soluble in alcohol and ether | |
| CC(S)=O | |
| 76.11 | |
| CHEBI:16555 | |
| 98% |
Chemical Identifiers
| 507-09-5 | |
| 76.11 | |
| DUYAAUVXQSMXQP-UHFFFAOYSA-N | |
| 10484 | |
| ethanethioic S-acid |
| C2H4OS | |
| MFCD00004853 | |
| thioacetic acid, ethanethioic acid, thiacetic acid, thioacetic s-acid, acetyl mercaptan, ethanethiolic acid, thiolacetic acid, thionoacetic acid, methanecarbothiolic acid, ch3cosh | |
| CHEBI:16555 | |
| CC(S)=O |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Toxic if swallowed.
Causes serious eye damage.
Highly flammable liquid and vapor.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER
GHS Signal Word: Danger
EINECSNumber : 208-063-8
RTECSNumber : AJ5600000
TSCA : TSCA
RUO – Research Use Only