missing translation for 'onlineSavingsMsg'
Learn More

Thioacetanilide 98.0+%, TCI America™

Catalog No. T01885G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T01885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T01885G Supplier TCI America Supplier No. T01885G

Chemical Identifiers

CAS 637-53-6
Molecular Formula C8H9NS
Molecular Weight (g/mol) 151.23
MDL Number MFCD00004942
InChI Key MWCGLTCRJJFXKR-UHFFFAOYSA-N
Synonym thioacetanilide, ethanethioamide, n-phenyl, n-phenylthioacetamide, acetanilide, thio, usaf ek-1902, unii-l9al2go03y, l9al2go03y, 1-phenylamino ethane-1-thione, acmc-209nhr
PubChem CID 820777
IUPAC Name N-phenylethanethioamide
SMILES CC(=S)NC1=CC=CC=C1

Specifications

Melting Point 76°C
Boiling Point 147°C
Quantity 5 g
Formula Weight 151.23
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Thioacetanilide
RTECSNumber AE7350000
TSCA Yes