missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thioacetamide, Crystal, BAKER ANALYZED™ Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
$9227.85 - $9227.85
Chemical Identifiers
| CAS | 62-55-5 |
|---|---|
| Molecular Formula | C2H5NS |
| Molecular Weight (g/mol) | 75.13 |
| MDL Number | MFCD00008070 |
| InChI Key | YUKQRDCYNOVPGJ-UHFFFAOYSA-N |
| Synonym | thioacetamide, acetothioamide, thiacetamide, acetamide, thio, thioacetimidic acid, acetimidic acid, thio, usaf cb-21, usaf ek-1719, rcra waste number u218, acetic acid, thiono-, amide |
| PubChem CID | 2723949 |
| ChEBI | CHEBI:32497 |
| IUPAC Name | ethanethioamide |
| SMILES | CC(N)=S |
Chemical Identifiers
| 62-55-5 | |
| 75.13 | |
| YUKQRDCYNOVPGJ-UHFFFAOYSA-N | |
| 2723949 | |
| ethanethioamide |
| C2H5NS | |
| MFCD00008070 | |
| thioacetamide, acetothioamide, thiacetamide, acetamide, thio, thioacetimidic acid, acetimidic acid, thio, usaf cb-21, usaf ek-1719, rcra waste number u218, acetic acid, thiono-, amide | |
| CHEBI:32497 | |
| CC(N)=S |
Specifications
| 62-55-5 | |
| C2H5NS | |
| 500 g | |
| YUKQRDCYNOVPGJ-UHFFFAOYSA-N | |
| ethanethioamide | |
| 2723949 | |
| 75.13 | |
| Crystals |
| Wide Mouth Amber Glass Bottle | |
| MFCD00008070 | |
| thioacetamide, acetothioamide, thiacetamide, acetamide, thio, thioacetimidic acid, acetimidic acid, thio, usaf cb-21, usaf ek-1719, rcra waste number u218, acetic acid, thiono-, amide | |
| CC(N)=S | |
| 75.13 | |
| CHEBI:32497 | |
| BAKER ANALYZED™ Reagent | |
| Thioacetamide |