missing translation for 'onlineSavingsMsg'
Learn More

Thioacetamide, 98%

Catalog No. AAA1292614
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1292614 25 g
AAA1292622 100 g
AAA1292636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1292614 Supplier Thermo Scientific Chemicals Supplier No. A1292614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 62-55-5 | C2H5NS | 75.13 g/mol

Thioacetamide is used in qualitative inorganic analysis as an in-situ source for sulfide ions. It is used in analytical chemistry as a source of hydrogen sulfide. It is an additive in enantioselective reduction of beta-keto esters with immobilized bakers yeast. It is also used as a vulcanizing agent of polymer and a crosslinking agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thioacetamide is used in qualitative inorganic analysis as an in-situ source for sulfide ions. It is used in analytical chemistry as a source of hydrogen sulfide. It is an additive in enantioselective reduction of beta-keto esters with immobilized bakers yeast. It is also used as a vulcanizing agent of polymer and a crosslinking agent.

Solubility
Soluble in ethanol, benzene and petroleum ether. Slightly soluble in water and ether.

Notes
Incompatible with strong oxidizing agents, strong acids and strong bases.

Chemical Identifiers

CAS 62-55-5
Molecular Formula C2H5NS
Molecular Weight (g/mol) 75.13
MDL Number MFCD00008070
InChI Key YUKQRDCYNOVPGJ-UHFFFAOYSA-N
Synonym thioacetamide, acetothioamide, thiacetamide, acetamide, thio, thioacetimidic acid, acetimidic acid, thio, usaf cb-21, usaf ek-1719, rcra waste number u218, acetic acid, thiono-, amide
PubChem CID 2723949
ChEBI CHEBI:32497
IUPAC Name ethanethioamide
SMILES CC(N)=S

Specifications

Melting Point 110°C to 114°C
Density 1.37
Assay Percent Range 98%
Quantity 25 g
Beilstein 506006
Merck Index 14,9319
Solubility Information Soluble in ethanol,benzene and petroleum ether; Slightly soluble in water and ether.
Formula Weight 75.13
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material Thioacetamide
Show More Show Less
Hazard Category H302-H315-H319-H336-H351-H373
Hazard Statement GHS H Statement
H350-H302-H315-H319
Precautionary Statement P201-P202-P260-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
EINECSNumber 200-541-4
RTECSNumber AC8925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only