Learn More
Greiner Bio-One ThinCert™ Tissue Culture Inserts
GREENER_CHOICE
Research tool for transport, co-culture, migration and invasion studies. Greiner Bio-One ThinCert™ Tissue Culture Inserts provides improved cell adhesion due to physical surface treatment.
$534.79 - $899.04
Products 14
Catalog Number Mfr. No. Pore Size No. of Wells Pore Density Price Quantity & Availability  
Catalog Number Mfr. No. Pore Size No. of Wells Pore Density Price Quantity & Availability  
07000647
View Documents Promotion Details
Greiner Bio-One
657630
3 μm 6 0.6 x 106 pores/cm2
Case of 24 Each for $534.79
Only null left
07000648
View Documents Promotion Details
Greiner Bio-One
657640
0.4 μm 6 1 x 108 pores/cm2
Case of 24 Each for $535.14
Only null left
07000676
View Documents Promotion Details
Greiner Bio-One
662631
3 μm 24 2 x 106 pores/cm2
Case of 48 Each for $769.61
Only null left
07000677
View Documents Promotion Details
Greiner Bio-One
662640
0.4 μm 24 1 x 108 pores/cm2
Case of 48 Each for $775.54
Only null left
07000690
View Documents Promotion Details
Greiner Bio-One
665641
0.4 μm 12 2 x 106 pores/cm2
Case of 48 Each for $889.32
Only null left
07000388
View Documents Promotion Details
Greiner Bio-One
665640
0.4 μm 12 1 x 108 pores/cm2
Case of 48 Each for $888.00
Only null left
07000389
View Documents Promotion Details
Greiner Bio-One
665610
1.0 μm 12 2 x 106 pores/cm2
Case of 48 Each for $899.04
Only null left
07000390
View Documents Promotion Details
Greiner Bio-One
665630
3.0 μm 12 0.6 x 106 pores/cm2
Case of 48 Each for $885.18
Only null left
07000391
GREENER_CHOICE
View Documents Promotion Details
Greiner Bio-One
665631
3.0 μm 12 2 x 106 pores/cm2
Case of 48 Each for $896.86
Only null left
07000392
View Documents Promotion Details
Greiner Bio-One
665638
8.0 μm 12 0.15 x 106 pores/cm2
Case of 48 Each for $824.50
Only null left
07000394
View Documents Promotion Details
Greiner Bio-One
662610
1.0 μm 24 2 x 106 pores/cm2
Case of 48 Each for $773.71
Only null left
07000395
View Documents Promotion Details
Greiner Bio-One
662630
3.0 μm 24 0.6 x 106 pores/cm2
Case of 48 Each for $777.00
Only null left
07000398
View Documents Promotion Details
Greiner Bio-One
657610
1.0 μm 6 2 x 106 pores/cm2
Case of 24 Each for $544.96
Only null left
07000399
View Documents Promotion Details
Greiner Bio-One
657631
3.0 μm 6 2 x 106 pores/cm2
Case of 24 Each for $613.25
Only null left
Description
Description
  • Clear polystyrene housing with sealed PET capillary pore membrane
  • Hanging geometry provides protection to lower compartment cell layer
  • Simplified pipetting due to self-lift geometry
  • Eccentric in-well positioning allows unequalled access to lower compartment
  • Available in 6, 12 and 24 well format