missing translation for 'onlineSavingsMsg'
Learn More

BRANDTECH™ BRAND™ 0.5 mL PCR Tubes with Attached Caps, BIO-CERT™ PCR QUALITY

Thin Wall PCR Tubes with attached caps ensure precise thermal transfer, RNase-/DNase-/DNA-free, and manufactured without lubricants or additives that could contaminate samples.

$299.94 - $337.50

Specifications

Certifications/Compliance RNase, DNase and DNA free
Volume (Metric) 0.5 mL
Quantity 1000 Pack
For Use With (Equipment) Thermal cyclers for excellent reproducible results in most PCR applications
Sterility Non-sterile
View More Specs

Products 5
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
1388259
View Documents Promotion Details
BRANDTECH™ BRAND™
781310
Clear
Pack of 1000 Each for $337.50
Only null left
 
1388260
View Documents Promotion Details
BRANDTECH™ BRAND™
781311
Rose
Pack of 1000 Each for $299.94
Only null left
 
1388262
View Documents Promotion Details
BRANDTECH™ BRAND™
781313
Green
Pack of 1000 Each for $299.94
Only null left
 
1388263
View Documents Promotion Details
BRANDTECH™ BRAND™
781314
Blue
Pack of 1000 Each for $305.22
Only null left
 
1388261
View Documents Promotion Details
BRANDTECH™ BRAND™
781312
Yellow
Pack of 1000 Each for $299.94
Only null left
 
Description

Description

BRAND ™ Disposable PCR Tubes are designed to deliver exceptional performance for PCR setups, small- and medium-throughput applications, and other workflows requiring precise thermal transfer and high cleanliness. Made from high-quality virgin polypropylene, these tubes ensure reliable results with features that enhance usability and sample integrity. Key features include:

  • BIO-CERT™ PCR QUALITY: Certified free of human DNA, RNase, DNase, pyrogens, and PCR inhibitors, ensuring the highest level of cleanliness for sensitive PCR applications.
  • Superior Heat Transfer: Extra thin-walled tubes optimize thermal transfer during PCR, reducing cycle times and providing consistent performance over a wide temperature range.
  • Reliable Sealing Caps: Tight-sealing caps prevent sample evaporation and loss due to heating, while offering easy-to-open and close functionality.
  • Color Options: Offered in multiple colors for simplified visual sample tracking and organization.
  • High-Quality Construction: Manufactured from high-quality virgin resins without lubricants or chemical additives that could contaminate reagents or samples.
  • Autoclavable: Designed to withstand autoclaving at 121°C (2 bar) in compliance with DIN EN 285.
Specifications

Specifications

RNase, DNase and DNA free
1000 Pack
Non-sterile
Flat Cap
Yes
Tube
Thin Wall PCR Tube
0.5 mL
Thermal cyclers for excellent reproducible results in most PCR applications
Polypropylene
Autoclavable
Single-use
0.5 mL
Videos
SDS
Documents

Documents