Learn More
BRANDTECH™ BRAND™ 0.5 mL PCR Tubes with Attached Caps, BIO-CERT™ PCR QUALITY
Thin Wall PCR Tubes with attached caps ensure precise thermal transfer, RNase-/DNase-/DNA-free, and manufactured without lubricants or additives that could contaminate samples.
Supplier: BRANDTECH™ BRAND™ 781311
Description
BRAND ™ Disposable PCR Tubes are designed to deliver exceptional performance for PCR setups, small- and medium-throughput applications, and other workflows requiring precise thermal transfer and high cleanliness. Made from high-quality virgin polypropylene, these tubes ensure reliable results with features that enhance usability and sample integrity. Key features include:
- BIO-CERT™ PCR QUALITY: Certified free of human DNA, RNase, DNase, pyrogens, and PCR inhibitors, ensuring the highest level of cleanliness for sensitive PCR applications.
- Superior Heat Transfer: Extra thin-walled tubes optimize thermal transfer during PCR, reducing cycle times and providing consistent performance over a wide temperature range.
- Reliable Sealing Caps: Tight-sealing caps prevent sample evaporation and loss due to heating, while offering easy-to-open and close functionality.
- Color Options: Offered in multiple colors for simplified visual sample tracking and organization.
- High-Quality Construction: Manufactured from high-quality virgin resins without lubricants or chemical additives that could contaminate reagents or samples.
- Autoclavable: Designed to withstand autoclaving at 121°C (2 bar) in compliance with DIN EN 285.
Chemical Identifiers
| RNase, DNase and DNA free | |
| 1000 Pack | |
| Non-sterile | |
| Flat Cap | |
| Yes | |
| Tube | |
| Thin Wall PCR Tube |
| 0.5 mL | |
| Thermal cyclers for excellent reproducible results in most PCR applications | |
| Polypropylene | |
| Autoclavable | |
| Single-use | |
| 0.5 mL |
Specifications
| RNase, DNase and DNA free | |
| 0.5 mL | |
| Thermal cyclers for excellent reproducible results in most PCR applications | |
| Polypropylene | |
| Autoclavable | |
| Single-use | |
| 0.5 mL |
| Rose | |
| 1000 Pack | |
| Non-sterile | |
| Flat Cap | |
| Yes | |
| Tube | |
| Thin Wall PCR Tube |