missing translation for 'onlineSavingsMsg'
Learn More

Thifluzamide 98.0+%, TCI America™

Catalog No. p-7137826
Change view
Click to view available options
Quantity:
250 mg
1 g
Catalog No. Quantity
T3648250MG 250 mg
T36481G 1 g
2 options

Catalog No. T3648250MG

Supplier: TCI America T3648250MG

Only null left

Chemical Identifiers

CAS 130000-40-7
Molecular Formula C13H6Br2F6N2O2S
Molecular Weight (g/mol) 528.061
MDL Number MFCD03428071
InChI Key WOSNCVAPUOFXEH-UHFFFAOYSA-N
Synonym N-[2,6-Dibromo-4-(trifluoromethoxy)phenyl]-2-methyl-4-(trifluoromethyl)thiazole-5-carboxamide
PubChem CID 86389
ChEBI CHEBI:81794
IUPAC Name N-[2,6-dibromo-4-(trifluoromethoxy)phenyl]-2-methyl-4-(trifluoromethyl)-1,3-thiazole-5-carboxamide
SMILES CC1=NC(=C(S1)C(=O)NC2=C(C=C(C=C2Br)OC(F)(F)F)Br)C(F)(F)F

Specifications

Melting Point 177°C
Quantity 250 mg
UN Number 3077
Formula Weight 528.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Thifluzamide