missing translation for 'onlineSavingsMsg'
Learn More

Thieno[3,2-d]pyrimidin-4(1H)-one 98.0+%, TCI America™

Catalog No. T33015G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T33015G 5 g
T33011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T33015G Supplier TCI America Supplier No. T33015G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16234-10-9
Molecular Formula C6H4N2OS
Molecular Weight (g/mol) 152.171
MDL Number MFCD00122140
InChI Key PZMKGWRBZNOIPQ-UHFFFAOYSA-N
PubChem CID 2760611
IUPAC Name 1H-thieno[3,2-d]pyrimidin-4-one
SMILES C1=CSC2=C1NC=NC2=O

Specifications

Melting Point 223°C
Color White
Quantity 5 g
Formula Weight 152.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Thieno[3,2-d]pyrimidin-4(1H)-one