accessibility menu, dialog, popup

UserName

CAS RN 16234-10-9

IUPAC Name: 1H,4H-thieno[3,2-d]pyrimidin-4-one
Synonyms: 1H-thieno[3,2-d]pyrimidin-4-one
Molecular Weight (g/mol): 152.17
Molecular Formula: C6H4N2OS
InChi Key: PZMKGWRBZNOIPQ-UHFFFAOYSA-N
SMILES: O=C1N=CNC2=C1SC=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results