missing translation for 'onlineSavingsMsg'
Learn More

Thieno[3,2-b]thiophene-2-carboxylic Acid 98.0+%, TCI America™

Catalog No. p-7137610
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3156200MG 200 mg
T31561G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3156200MG Supplier TCI America Supplier No. T3156200MG
Only null left

Chemical Identifiers

CAS 1723-27-9
Molecular Formula C7H4O2S2
Molecular Weight (g/mol) 184.23
MDL Number MFCD00115174
InChI Key GVZXSZWCZGKLRS-UHFFFAOYSA-N
Synonym thieno 3,2-b thiophene-2-carboxylic acid, thieno 3,2-b thiophene-5-carboxylic acid, thieno 3,2-b thiophene-2-carboxylicacid, thiopheno 3,2-b thiophene-2-carboxylic acid, zlchem 802, maybridge3_006605, thienothiophene carboxylic acid, 5-thieno 3,2-b thiophenecarboxylic acid, nuk
PubChem CID 660319
IUPAC Name thieno[3,2-b]thiophene-2-carboxylic acid
SMILES OC(=O)C1=CC2=C(S1)C=CS2

Specifications

Melting Point 223°C
Color Yellow
Quantity 200 mg
Formula Weight 184.23
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Thieno[3,2-b]thiophene-2-carboxylic Acid