accessibility menu, dialog, popup

UserName

CAS RN 1723-27-9

IUPAC Name: thieno[3,2-b]thiophene-2-carboxylic acid
Synonyms: thieno[3,2-b]thiophene-2-carboxylic acid
Molecular Weight (g/mol): 184.23
Molecular Formula: C7H4O2S2
InChi Key: GVZXSZWCZGKLRS-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC2=C(S1)C=CS2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results