missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thieno[3,2-b]thiophene-2,5-dicarboxaldehyde 93.0+%, TCI America™
$167.76 - $551.76
Chemical Identifiers
| CAS | 37882-75-0 |
|---|---|
| Molecular Formula | C8H4O2S2 |
| Molecular Weight (g/mol) | 196.24 |
| MDL Number | MFCD03931117 |
| InChI Key | UBKNFAFBINVTOP-UHFFFAOYSA-N |
| PubChem CID | 361865 |
| IUPAC Name | thieno[3,2-b]thiophene-2,5-dicarbaldehyde |
| SMILES | O=CC1=CC2=C(S1)C=C(S2)C=O |
Chemical Identifiers
| 37882-75-0 | |
| 196.24 | |
| UBKNFAFBINVTOP-UHFFFAOYSA-N | |
| thieno[3,2-b]thiophene-2,5-dicarbaldehyde |
| C8H4O2S2 | |
| MFCD03931117 | |
| 361865 | |
| O=CC1=CC2=C(S1)C=C(S2)C=O |
Specifications
| 37882-75-0 | |
| C8H4O2S2 | |
| 200 mg | |
| O=CC1=CC2=C(S1)C=C(S2)C=O | |
| 196.24 | |
| 196.24 | |
| Crystalline Powder |
| 274°C | |
| MFCD03931117 | |
| UBKNFAFBINVTOP-UHFFFAOYSA-N | |
| thieno[3,2-b]thiophene-2,5-dicarbaldehyde | |
| 361865 | |
| ≥93.0% (GC) | |
| Thieno[3,2-b]thiophene-2,5-dicarboxaldehyde |
Safety and Handling
TSCA : No