accessibility menu, dialog, popup

UserName

CAS RN 37882-75-0

IUPAC Name: thieno[3,2-b]thiophene-2,5-dicarbaldehyde
Synonyms: thieno[3,2-b]thiophene-2,5-dicarbaldehyde
Molecular Weight (g/mol): 196.24
Molecular Formula: C8H4O2S2
InChi Key: UBKNFAFBINVTOP-UHFFFAOYSA-N
SMILES: O=CC1=CC2=C(S1)C=C(S2)C=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results