missing translation for 'onlineSavingsMsg'
Learn More

Thieno[3,2-b]thiophene-2,5-dicarboxaldehyde 93.0+%, TCI America™

Catalog No. T3212200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3212200MG 200 mg
T32121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3212200MG Supplier TCI America Supplier No. T3212200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 37882-75-0
Molecular Formula C8H4O2S2
Molecular Weight (g/mol) 196.24
MDL Number MFCD03931117
InChI Key UBKNFAFBINVTOP-UHFFFAOYSA-N
PubChem CID 361865
IUPAC Name thieno[3,2-b]thiophene-2,5-dicarbaldehyde
SMILES O=CC1=CC2=C(S1)C=C(S2)C=O

Specifications

Melting Point 274°C
Quantity 200 mg
Formula Weight 196.24
Percent Purity ≥93.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Thieno[3,2-b]thiophene-2,5-dicarboxaldehyde