missing translation for 'onlineSavingsMsg'
Learn More

Thieno[3,2-b]pyridin-7-ol, 96%

Catalog No. AAH5017203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5017203 1 g
AAH5017206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5017203 Supplier Thermo Scientific Chemicals Supplier No. H5017203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 107818-20-2 | C7H5NOS | 151.183 g/mol

Thieno[3,2-b]pyridin-7-ol is used for pharmaceutical intermediates, as drugs, food, cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thieno[3,2-b]pyridin-7-ol is used for pharmaceutical intermediates, as drugs, food, cosmetics.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 107818-20-2
Molecular Formula C7H5NOS
Molecular Weight (g/mol) 151.183
MDL Number MFCD01863636
InChI Key AACVULYSNJAKEQ-UHFFFAOYSA-N
Synonym thieno 3,2-b pyridin-7-ol, thieno 3,2-b pyridin-7 4h-one, 7-hydroxythieno 3,2-b pyridine, 4h-thieno 3,2-b pyridin-7-one, thiopheno 3,2-b pyridin-7-ol, thieno 3 2-b pyridin-7-ol, pubchem22356, acmc-2098xl, ksc173k8p
PubChem CID 5074765
IUPAC Name 4H-thieno[3,2-b]pyridin-7-one
SMILES C1=CNC2=C(C1=O)SC=C2

Specifications

Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 151.19
Percent Purity 96%
Chemical Name or Material Thieno[3,2-b]pyridin-7-ol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only