accessibility menu, dialog, popup

UserName

CAS RN 107818-20-2

IUPAC Name: 4H,7H-thieno[3,2-b]pyridin-7-one
Synonyms: 4H-thieno[3,2-b]pyridin-7-one
Molecular Weight (g/mol): 151.18
Molecular Formula: C7H5NOS
InChi Key: AACVULYSNJAKEQ-UHFFFAOYSA-N
SMILES: O=C1C=CNC2=C1SC=C2
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)