missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Thidiazuron

Catalog No. AAJ66663MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66663MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ66663MD Supplier Thermo Scientific Chemicals Supplier No. J66663MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

220.25 g/mol

Thidiazuron is a plant growth regulator used in micropropagations to supplement media such as Murashige and Skoog medium. TDZ promotes plant organogenesis and plant regeneration. Thidiazuron enables the production of virus-free stock plants, speeds up breeding, and provides new genotypes for the market.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Molecular Weight (g/mol) 220.25
Quantity 250 mg
Packaging Plastic bottle
Form Powder

RUO – Research Use Only