missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Thiazolyl Blue tetrazolium bromide, 98%

Catalog No. AAL1193903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1193903 1 g
AAL1193906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1193903 Supplier Thermo Scientific Chemicals Supplier No. L1193903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 298-93-1 | C18H16BrN5S | 414.33 g/mol

Used in measurement of cell proliferation. Produces a yellowish solution that is converted to dark blue, water-insoluble MTT formazan by mitochondrial dehydrogenases of living cells.Thiazolyl Blue tetrazolium bromide is a dye used in measurement of cell proliferation. It is used as a direct indicator of cytotoxicity (such as for screening cancer drugs) and apoptosis. As an electron acceptor, it is utilized for studying nicotinamide adenine dinucleotide phosphate (NADP)-dependent dehydrogenases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in measurement of cell proliferation. Produces a yellowish solution that is converted to dark blue, water-insoluble MTT formazan by mitochondrial dehydrogenases of living cells.Thiazolyl Blue tetrazolium bromide is a dye used in measurement of cell proliferation. It is used as a direct indicator of cytotoxicity (such as for screening cancer drugs) and apoptosis. As an electron acceptor, it is utilized for studying nicotinamide adenine dinucleotide phosphate (NADP)-dependent dehydrogenases.

Solubility
Soluble in water, ethanol, 2-methoxyethanol and phosphate buffered saline.

Notes
Light, air and moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 298-93-1
Molecular Formula C18H16BrN5S
Molecular Weight (g/mol) 414.33
MDL Number MFCD00011964,MFCD00066662
InChI Key AZKSAVLVSZKNRD-UHFFFAOYSA-M
Synonym thiazolyl blue, thiazolyl blue tetrazolium bromide, mmt tetrazolium, methylthiazoletetrazolium, mtt, thiazolyl blue monotetrazolium, thiazole blue, 3-4,5-dimethylthiazol-2-yl-2,5-diphenyltetrazolium bromide, mtt van, unii-euy85h477i
PubChem CID 64965
ChEBI CHEBI:53233
IUPAC Name 2-(3,5-diphenyltetrazol-2-ium-2-yl)-4,5-dimethyl-1,3-thiazole;bromide
SMILES [Br-].CC1=C(C)N=C(S1)[N+]1=NC(=NN1C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point ∽195°C (decomposition)
Odor Odorless
Quantity 1 g
Beilstein 3825277
Sensitivity Air, light, and moisture sensitive
Solubility Information Soluble in water,ethanol,2-methoxyethanol and phosphate buffered saline.
Formula Weight 414.33
Percent Purity 98%
Chemical Name or Material Thiazolyl Blue tetrazolium bromide
Hazard Category H315-H319-H335-H341
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
EINECSNumber 206-069-5
RTECSNumber XF8060000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only