accessibility menu, dialog, popup

UserName

BrCH3SNH3CNNNN

CAS RN 298-93-1

IUPAC Name: 2-(4,5-dimethyl-1,3-thiazol-2-yl)-3,5-diphenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium bromide
Synonyms: thiazolyl blue methylthiazoletetrazolium mmt tetrazolium MTT thiazolyl blue monotetrazolium thiazolyl blue tetrazolium bromide tmchb
Molecular Weight (g/mol): 414.33
Molecular Formula: C18H16BrN5S
InChi Key: AZKSAVLVSZKNRD-UHFFFAOYSA-M
SMILES: [Br-].CC1=C(C)N=C(S1)[N+]1=NC(=NN1C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (4)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (4)
Physical Form 
  • (2)
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)