missing translation for 'onlineSavingsMsg'
Learn More

Thiazolidine-2-carboxylic acid, 97%

Catalog No. AAA1156406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1156406 5 g
AAA1156414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1156406 Supplier Thermo Scientific Chemicals Supplier No. A1156406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 16310-13-7 | C4H7NO2S | 133.165 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16310-13-7
Molecular Formula C4H7NO2S
Molecular Weight (g/mol) 133.165
MDL Number MFCD00068005
InChI Key ULSZVNJBVJWEJE-UHFFFAOYSA-N
Synonym thiazolidine-2-carboxylic acid, 1,3-thiazolane-2-carboxylic acid, 2-thiazolidinecarboxylic acid, beta-thiaproline, thiazolidine-2-carboxylicacid, acmc-20aiwa, thiazolidine carboxylic acid, 2-triazolidinecarboxylic acid, ulszvnjbvjweje-uhfffaoysa, tetrahydrothiazole-2-carboxylic acid
PubChem CID 42486
IUPAC Name 1,3-thiazolidine-2-carboxylic acid
SMILES C1CSC(N1)C(=O)O

Specifications

Melting Point ∼184°C (decomposition)
Quantity 5 g
Beilstein 112783
Formula Weight 133.17
Percent Purity 97%
Chemical Name or Material Thiazolidine-2-carboxylic acid
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only