missing translation for 'onlineSavingsMsg'
Learn More

Thiazolidine-2,4-dicarboxylic Acid 98.0+%, TCI America™

Catalog No. t13255g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T13255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T13255G Supplier TCI America Supplier No. T13255G
Only null left

Chemical Identifiers

CAS 30097-06-4
Molecular Formula C5H7NO4S
Molecular Weight (g/mol) 177.174
MDL Number MFCD00145399
InChI Key DAXBISKSIDBYEU-UHFFFAOYSA-N
Synonym tidiacic, thiazolidine-2,4-dicarboxylic acid, 2,4-thiazolidinedicarboxylic acid, 1,3-thiazolane-2,4-dicarboxylic acid, tdca, tidiacicum, tidiacico, 2,4-thiazolidinedicarboxylic acid, trans, tidiacic inn:dcf, tidiacicum inn-latin
PubChem CID 72150
IUPAC Name 1,3-thiazolidine-2,4-dicarboxylic acid
SMILES C1C(NC(S1)C(=O)O)C(=O)O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 177.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Thiazolidine-2,4-dicarboxylic Acid