missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thiazole 98.0+%, TCI America™
$48.46 - $190.13
Chemical Identifiers
| CAS | 288-47-1 |
|---|---|
| Molecular Formula | C3H3NS |
| Molecular Weight (g/mol) | 85.12 |
| MDL Number | MFCD00005315 |
| InChI Key | FZWLAAWBMGSTSO-UHFFFAOYSA-N |
| Synonym | thiazole, thiazol, unii-320rcw8pef, fema no. 3615, ccris 3205, 320rcw8pef, lom, racemic thiazole, 5h-thiazol-1-ium |
| PubChem CID | 9256 |
| ChEBI | CHEBI:43732 |
| IUPAC Name | 1,3-thiazole |
| SMILES | S1C=CN=C1 |
Chemical Identifiers
| 288-47-1 | |
| 85.12 | |
| FZWLAAWBMGSTSO-UHFFFAOYSA-N | |
| 9256 | |
| 1,3-thiazole |
| C3H3NS | |
| MFCD00005315 | |
| thiazole, thiazol, unii-320rcw8pef, fema no. 3615, ccris 3205, 320rcw8pef, lom, racemic thiazole, 5h-thiazol-1-ium | |
| CHEBI:43732 | |
| S1C=CN=C1 |
Specifications
| 288-47-1 | |
| Yellow | |
| C3H3NS | |
| 5 g | |
| thiazole, thiazol, unii-320rcw8pef, fema no. 3615, ccris 3205, 320rcw8pef, lom, racemic thiazole, 5h-thiazol-1-ium | |
| S1C=CN=C1 | |
| 85.12 | |
| CHEBI:43732 | |
| ≥98.0% (GC) | |
| Thiazole |
| -33°C | |
| 118°C | |
| MFCD00005315 | |
| 1993 | |
| FZWLAAWBMGSTSO-UHFFFAOYSA-N | |
| 1,3-thiazole | |
| 9256 | |
| 85.12 | |
| Liquid |
Safety and Handling
RTECSNumber : XJ1290000
TSCA : Yes