missing translation for 'onlineSavingsMsg'
Learn More

Thiazole, 99%

Catalog No. AC166130010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC166130010 1 g Glass bottle
AC166130050 5 g Glass bottle
AC166130250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC166130010 Supplier Thermo Scientific Chemicals Supplier No. 166130010

CAS: 288-47-1 | C3H3NS | 85.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 288-47-1
Molecular Formula C3H3NS
Molecular Weight (g/mol) 85.12
MDL Number MFCD00005315
InChI Key FZWLAAWBMGSTSO-UHFFFAOYSA-N
Synonym thiazole, thiazol, unii-320rcw8pef, fema no. 3615, ccris 3205, 320rcw8pef, lom, racemic thiazole, 5h-thiazol-1-ium
PubChem CID 9256
ChEBI CHEBI:43732
IUPAC Name 1,3-thiazole
SMILES S1C=CN=C1

Specifications

Color Colorless to Yellow
Density 1.1980g/mL
Boiling Point 117.0°C to 118.0°C
Flash Point 26°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5365 to 1.5395
Quantity 1 g
Beilstein 27, 15
Specific Gravity 1.198
Merck Index 15, 9459
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol and ether
Formula Weight 85.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Thiazole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
EINECSNumber 206-021-3

RUO – Research Use Only