Learn More
Thiazole, 99%
CAS: 288-47-1 | C3H3NS | 85.12 g/mol
$70.97 - $766.14
Chemical Identifiers
| CAS | 288-47-1 |
|---|---|
| Molecular Formula | C3H3NS |
| Molecular Weight (g/mol) | 85.12 |
| MDL Number | MFCD00005315 |
| InChI Key | FZWLAAWBMGSTSO-UHFFFAOYSA-N |
| Synonym | thiazole, thiazol, unii-320rcw8pef, fema no. 3615, ccris 3205, 320rcw8pef, lom, racemic thiazole, 5h-thiazol-1-ium |
| PubChem CID | 9256 |
| ChEBI | CHEBI:43732 |
| IUPAC Name | 1,3-thiazole |
| SMILES | S1C=CN=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0997003
|
Thermo Scientific Chemicals
L0997003 |
1 g |
Each for $70.97
|
|
|||||
|
AAL0997014
|
Thermo Scientific Chemicals
L0997014 |
25 g |
Each for $766.14
|
|
|||||
Description
Thiazole is used as a flavoring agent and in the preparation of dyes and rubber accelerators. It serves as a component of the vitamin thiamine (B1). It acts as a protected formyl group used in natural product synthesis. It reacts with alkyl lithium and Grignardâs reagent to prepare organometallic complexes. It is involved in the electrophilic aromatic substitution and nucleophilic aromatic substitution at C-5 and C-2 positions respectively. Further, it undergoes alkylation reaction to get thiazolium cation, which is used as a catalyst in the Stetter reaction and the Benzoin condensation. In addition to this, it is involved in the preparation of alagebrium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsThiazole is used as a flavoring agent and in the preparation of dyes and rubber accelerators. It serves as a component of the vitamin thiamine (B1). It acts as a protected formyl group used in natural product synthesis. It reacts with alkyl lithium and Grignardâs reagent to prepare organometallic complexes. It is involved in the electrophilic aromatic substitution and nucleophilic aromatic substitution at C-5 and C-2 positions respectively. Further, it undergoes alkylation reaction to get thiazolium cation, which is used as a catalyst in the Stetter reaction and the Benzoin condensation. In addition to this, it is involved in the preparation of alagebrium.
Solubility
Soluble in ethanol and ether. Slightly soluble in water.
Notes
Air and light sensitive. Incompatible with acids, acid chlorides, acid anhydrides, strong oxidizing agents and carbon dioxide.
Chemical Identifiers
| 288-47-1 | |
| 85.12 | |
| FZWLAAWBMGSTSO-UHFFFAOYSA-N | |
| 9256 | |
| 1,3-thiazole |
| C3H3NS | |
| MFCD00005315 | |
| thiazole, thiazol, unii-320rcw8pef, fema no. 3615, ccris 3205, 320rcw8pef, lom, racemic thiazole, 5h-thiazol-1-ium | |
| CHEBI:43732 | |
| S1C=CN=C1 |
Specifications
| 288-47-1 | |
| 1.2 | |
| 22°C (71°F) | |
| C3H3NS | |
| MFCD00005315 | |
| UN1993 | |
| Air and light sensitive | |
| thiazole, thiazol, unii-320rcw8pef, fema no. 3615, ccris 3205, 320rcw8pef, lom, racemic thiazole, 5h-thiazol-1-ium | |
| FZWLAAWBMGSTSO-UHFFFAOYSA-N | |
| 1,3-thiazole | |
| 9256 | |
| 85.13 | |
| Thiazole |
| -33°C | |
| 117°C to 118°C | |
| Unpleasant | |
| 1.539 | |
| 1 g | |
| 103852 | |
| 14,9307 | |
| Soluble in ethanol and ether. Slightly soluble in water. | |
| S1C=CN=C1 | |
| 85.12 | |
| CHEBI:43732 | |
| 99% |
Safety and Handling
GHS H Statement
H225-H302-H319
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes serious eye irritation.
P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P330-P370+P378q-P501c
H226-H302
DOTInformation : Hazard Class: 3; Packaging Group: III
EINECSNumber : 206-021-3
RTECSNumber : XJ1290000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only