missing translation for 'onlineSavingsMsg'
Learn More

Thiazole-2-carboxylic acid, 95%

CAS: 14190-59-1 | C4H3NO2S | 129.133 g/mol

Supplier:  Thermo Scientific Chemicals H6362003

 View more versions of this product

Catalog No. AAH6362003


Only null left
Explore available promotions

Description

Description

Thiazole-2-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiazole-2-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
Specifications

Specifications

Thiazole-2-carboxylic acid
C4H3NO2S
1 g
Slightly soluble in water.
C1=CSC(=N1)C(=O)O
129.133
129.14
Solid
14190-59-1
MFCD02094169
thiazole-2-carboxylic acid, 2-thiazolecarboxylic acid, thiazole-2-carboxylicacid, 2-carboxy-1,3-thiazole, thiazolecarboxylic acid, pubchem16257, thiazole carboxylic acid, 2-carboxythiazole, acmc-209cn5
IJVLVRYLIMQVDD-UHFFFAOYSA-N
1,3-thiazole-2-carboxylic acid
2762733
95%

Chemical Identifiers

14190-59-1
129.133
IJVLVRYLIMQVDD-UHFFFAOYSA-N
2762733
C1=CSC(=N1)C(=O)O
C4H3NO2S
MFCD02094169
thiazole-2-carboxylic acid, 2-thiazolecarboxylic acid, thiazole-2-carboxylicacid, 2-carboxy-1,3-thiazole, thiazolecarboxylic acid, pubchem16257, thiazole carboxylic acid, 2-carboxythiazole, acmc-209cn5
1,3-thiazole-2-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only