missing translation for 'onlineSavingsMsg'
Learn More

Thiazole-2-carboxylic acid, 95%

Catalog No. p-7055120
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63620MD 250 mg
AAH6362003 1 g
AAH6362006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH63620MD Supplier Thermo Scientific Chemicals Supplier No. H63620MD
Only null left

CAS: 14190-59-1 | C4H3NO2S | 129.133 g/mol

Thiazole-2-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiazole-2-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.

Chemical Identifiers

CAS 14190-59-1
Molecular Formula C4H3NO2S
Molecular Weight (g/mol) 129.133
MDL Number MFCD02094169
InChI Key IJVLVRYLIMQVDD-UHFFFAOYSA-N
Synonym thiazole-2-carboxylic acid, 2-thiazolecarboxylic acid, thiazole-2-carboxylicacid, 2-carboxy-1,3-thiazole, thiazolecarboxylic acid, pubchem16257, thiazole carboxylic acid, 2-carboxythiazole, acmc-209cn5
PubChem CID 2762733
IUPAC Name 1,3-thiazole-2-carboxylic acid
SMILES C1=CSC(=N1)C(=O)O

Specifications

Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 129.14
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Thiazole-2-carboxylic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only