missing translation for 'onlineSavingsMsg'
Learn More

Thiazole-2-carboxaldehyde, 95%

Catalog No. AAH6332903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6332903 1 g
AAH6332906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6332903 Supplier Thermo Scientific Chemicals Supplier No. H6332903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10200-59-6 | C4H3NOS | 113.134 g/mol

Thiazole-2-carboxaldehyde may be used as a reactant in the following syntheses Benzothiazine N-acylhydrazones, having potential antinociceptive and anti-inflammatory activity, Thiazole-2-yl-(amino)methylphosphonate diethyl esters, Imino ester by reacting with L-leucine t-butyl ester hydrochloride. Also used in building block for taxane analogs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiazole-2-carboxaldehyde may be used as a reactant in the following syntheses Benzothiazine N-acylhydrazones, having potential antinociceptive and anti-inflammatory activity, Thiazole-2-yl-(amino)methylphosphonate diethyl esters, Imino ester by reacting with L-leucine t-butyl ester hydrochloride. Also used in building block for taxane analogs.

Solubility
Fully miscible with water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.

Chemical Identifiers

CAS 10200-59-6
Molecular Formula C4H3NOS
Molecular Weight (g/mol) 113.134
MDL Number MFCD00142924
InChI Key ZGTFNNUASMWGTM-UHFFFAOYSA-N
Synonym 2-thiazolecarboxaldehyde, thiazole-2-carbaldehyde, 2-formylthiazole, thiazole-2-carboxaldehyde, 2-formyl thiazole, thiazolecarboxaldehyde, 2-formyl-1,3-thiazole, 1,3-thiazole-2-carboxaldehyde, 2-thiazolecarbaldehyde, pubchem9929
PubChem CID 2734903
ChEBI CHEBI:43623
IUPAC Name 1,3-thiazole-2-carbaldehyde
SMILES C1=CSC(=N1)C=O

Specifications

Color Colorless to Yellow
Density 1.288 g/mL
Boiling Point 61°C to 63°C (15 mmHg)
Flash Point 68°C (154°F)
Refractive Index 1.574
Quantity 1 g
Solubility Information Fully miscible with water.
Formula Weight 113.13
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Thiazole-2-carboxaldehyde
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only