accessibility menu, dialog, popup

UserName

OSN

CAS RN 10200-59-6

IUPAC Name: 1,3-thiazole-2-carbaldehyde
Synonyms: 1,3-thiazole-2-carbaldehyde
Molecular Weight (g/mol): 113.13
Molecular Formula: C4H3NOS
InChi Key: ZGTFNNUASMWGTM-UHFFFAOYSA-N
SMILES: O=CC1=NC=CS1
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (6)
  • (3)
  • (3)