missing translation for 'onlineSavingsMsg'
Learn More

Thiamine nitrate

Catalog No. p-7041720
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6001409 10 g
AAJ6001414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6001409 Supplier Thermo Scientific Chemicals Supplier No. J6001409

CAS: 532-43-4 | C12H17N5O4S | 327.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 532-43-4
Molecular Formula C12H17N5O4S
Molecular Weight (g/mol) 327.36
MDL Number MFCD00036330
InChI Key UIERGBJEBXXIGO-UHFFFAOYSA-N
Synonym thiamine nitrate, thiamine mononitrate, vitamin b1 nitrate, vitamin b1 mononitrate, aneurine mononitrate, aneurine nitrate, betabion mononitrate, thiamine nitrate salt, vitamin b sub 1 nitrate, thiamine mononitrate usp
PubChem CID 10762
IUPAC Name 2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethanol;nitrate
SMILES [O-][N+]([O-])=O.CC1=C(CCO)[S+]=CN1CC1=CN=C(C)N=C1N

Specifications

Melting Point 190°C
Color White
Quantity 10 g
Merck Index 14,9295
Formula Weight 327.36
Physical Form Powder
Chemical Name or Material Thiamine nitrate
EINECSNumber 208-537-4
RTECSNumber XI7400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only