missing translation for 'onlineSavingsMsg'
Learn More

Thiamine Mononitrate, FCC, 98-102%, Spectrum™ Chemical

Catalog No. p-7158379
Change view
Click to view available options
Quantity:
500 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18607363 500 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18607363 Supplier Spectrum Chemical Mfg Cor Supplier No. T1054500GM
Only null left

T1054, 532-43-4, C12H17N5O4S

Spectrum™ Chemical Thiamine Mononitrate, FCC is in the B vitamin group and is water soluble. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 532-43-4
Molecular Formula C12H17N5O4S
Molecular Weight (g/mol) 327.36
MDL Number MFCD00036330
InChI Key UIERGBJEBXXIGO-UHFFFAOYSA-N
IUPAC Name 3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-5-(2-hydroxyethyl)-4-methyl-3H-1λ⁴,3-thiazol-1-ylium nitrate
SMILES [O-][N+]([O-])=O.CC1=C(CCO)[S+]=CN1CC1=CN=C(C)N=C1N

Specifications

CAS Max % 1
pH 6.0 to 7.5
Ignition Residue 0.002
Loss on Drying 0.01
Packaging Amber Glass Bottle
Quantity 500 g
Percent Purity 98 to 102%
Grade FCC