missing translation for 'onlineSavingsMsg'
Learn More

Thiamine Impurity E Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013848
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013848 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013848 Supplier Merck Emd Millipore Supplier No. PHR175920MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thiamine Impurity E is an impurity of B class vitamins such as thiamine. Thiamine is important for primary metabolism for all living organisms and is also used in the treatment or prevention of beriberi.

Specifications

CAS 299-35-4
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C12H16N4 OS2
Synonym 3-[(4-Amino-2-methylpyrimidin-5-yl)methyl]-5-(2-hydroxyethyl)-4-methylthiazol-2(3 H)-thione; Thioxothiamine
Type Certified Reference Material
Quantity 20 mg
Formula Weight 296.41
Grade Certified Reference Material