missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thiamine Hydrochloride, FCC, 98-102%, Spectrum™ Chemical
T1053, 67-03-8, C12H17ClN4OS·HCl
Supplier: Spectrum Chemical Mfg Cor T1053100GM
Description
Spectrum™ Chemical Thiamine Hydrochloride, FCC is the hydrochloride salt of vitamin B1 and used to treat or prevent vitamin B1 deficiency. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 67-03-8 | |
| 2.7 to 3.4 | |
| Amber Glass Bottle | |
| 100 g | |
| [H+].[Cl-].[Cl-].CC1=C(CCO)SC=[N+]1CC1=CN=C(C)N=C1N | |
| 337.26 | |
| FCC |
| 1 | |
| 0.002 | |
| C12H18Cl2N4OS | |
| DPJRMOMPQZCRJU-UHFFFAOYSA-M | |
| hydrogen 3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-5-(2-hydroxyethyl)-4-methyl-1,3-thiazol-3-ium dichloride | |
| 98 to 102% |
Chemical Identifiers
| 67-03-8 | |
| 337.26 | |
| hydrogen 3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-5-(2-hydroxyethyl)-4-methyl-1,3-thiazol-3-ium dichloride |
| C12H18Cl2N4OS | |
| DPJRMOMPQZCRJU-UHFFFAOYSA-M | |
| [H+].[Cl-].[Cl-].CC1=C(CCO)SC=[N+]1CC1=CN=C(C)N=C1N |