missing translation for 'onlineSavingsMsg'
Learn More

Thiamine Hydrochloride, FCC, 98-102%, Spectrum™ Chemical

Catalog No. p-7158377
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18607351 25 g Amber Glass Bottle
18607352 100 g Amber Glass Bottle
18607353 500 g Amber Glass Bottle
18607354 1 kg Poly Pail
18607355 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18607351 Supplier Spectrum Chemical Mfg Cor Supplier No. T105325GM
Only null left

T1053, 67-03-8, C12H17ClN4OS·HCl

Spectrum™ Chemical Thiamine Hydrochloride, FCC is the hydrochloride salt of vitamin B1 and used to treat or prevent vitamin B1 deficiency. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 67-03-8
Molecular Formula C12H18Cl2N4OS
Molecular Weight (g/mol) 337.26
InChI Key DPJRMOMPQZCRJU-UHFFFAOYSA-M
IUPAC Name hydrogen 3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-5-(2-hydroxyethyl)-4-methyl-1,3-thiazol-3-ium dichloride
SMILES [H+].[Cl-].[Cl-].CC1=C(CCO)SC=[N+]1CC1=CN=C(C)N=C1N

Specifications

CAS Max % 1
pH 2.7 to 3.4
Ignition Residue 0.002
Packaging Amber Glass Bottle
Quantity 25 g
Percent Purity 98 to 102%
Grade FCC