missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Thiacloprid

Catalog No. AC464392500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464392500 250 mg
AC464390010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC464392500 Supplier Thermo Scientific Chemicals Supplier No. 464392500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 111988-49-9 | C10H9ClN4S | 252.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 111988-49-9
Molecular Formula C10H9ClN4S
Molecular Weight (g/mol) 252.72
InChI Key HOKKPVIRMVDYPB-UHFFFAOYSA-N
IUPAC Name ({3-[(6-chloropyridin-3-yl)methyl]-1,3-thiazolidin-2-ylidene}amino)formonitrile
SMILES ClC1=CC=C(CN2CCSC2=NC#N)C=N1

Specifications

Melting Point 133.0°C to 136.0°C
Quantity 250 mg
Solubility Information Solubility aq. soln.: .. Other solubilities: insoluble in chloroform & methanol
Formula Weight 252.72
Chemical Name or Material Thiacloprid
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Toxic if swallowed.

Harmful if inhaled.

May cause drowsiness or dizziness.

Suspected of causing cancer.

May damage fertility.

May damage the unborn child.

Very toxic to aquatic life with long last
Precautionary Statement GHS P Statement Wash face, hands and any exposed skin thoroughly after handling.

IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only