accessibility menu, dialog, popup

UserName

ClNSNNN(E/Z)

CAS RN 111988-49-9

IUPAC Name: ({3-[(6-chloropyridin-3-yl)methyl]-1,3-thiazolidin-2-ylidene}amino)formonitrile
Synonyms: thiacloprid calypso calypso 70WG
Molecular Weight (g/mol): 252.72
Molecular Formula: C10H9ClN4S
InChi Key: HOKKPVIRMVDYPB-UHFFFAOYSA-N
SMILES: ClC1=CC=C(CN2CCSC2=NC#N)C=N1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)