missing translation for 'onlineSavingsMsg'
Learn More

Thiabendazole, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111019560
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019560 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019560 Supplier Merck Emd Millipore Supplier No. PHR1259500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thiabendazole is an orally effective broad spectrum anthelmintic and antimycotic drug with limited side-effects.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specifications

CAS 148-79-8
Physical Form Neat
Linear Formula C10H7N3S
MDL Number MFCD00005587
Synonym Thiabendazole; 2-(4-Thiazolyl)benzimidazole
Quantity 500 mg
Formula Weight 201.25
Grade Certified Reference Material