missing translation for 'onlineSavingsMsg'
Learn More

Thiabendazole, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111019214
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019214 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019214 Supplier Merck Emd Millipore Supplier No. 6755450MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.

Specifications

CAS 148-79-8
Product Line TraceCERT
Linear Formula C10H7N3S
MDL Number MFCD00005587
Synonym 2-(4-Thiazolyl)benzimidazole
Quantity 50 mg
Formula Weight 201.25
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC