Learn More
Sonoco™ ThermoSafe Insulated EPS with Corrugated Containers and Coolers (Medium)
Large selection of cost-effective, reusable EPS foam in corrugated stock containers available in small, medium, and large sizes. EPS foam with corrugated shippers for the safe storage and delivery of most refrigerated or frozen products.
$228.24 - $600.94
Products 11
Catalog Number Mfr. No. Dimensions (L x W x H) Exterior Dimensions (D x W x H) Interior Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W x H) Exterior Dimensions (D x W x H) Interior Price Quantity & Availability  
0353055
View Documents Promotion Details
Sonoco™
325UPS
14.88 X 13.25 X 10.13 in. 11.63 X 9.88 X 7 in.
Case of 6 Each for $468.91
Only null left
035291
View Documents Promotion Details
Sonoco™
326
14.88 X 13.13 X 12.13 in. 11.75 X 10 X 8.63 in.
Case of 6 Each for $262.79
Only null left
0353017
View Documents Promotion Details
Sonoco™
TS355
14 X 14 X 14.63 in. 11 X 11 X 11.63 in.
Pack of 4 Each for $406.85
Only null left
0353043
View Documents Promotion Details
Sonoco™
TS453
16 X 16 X 15.5 in. 12 X 12 X 11.5 in.
Case of 3 Each for $431.48
Only null left
0352827
View Documents Promotion Details
Sonoco™
TS479
15.5 X 11.5 X 8 in. 13.13 X 9.25 X 6 in.
Case of 8 Each for $600.94
Only null left
03001104
View Documents Promotion Details
Sonoco™
611
15.5 X 15.5 X 10 in. 12.5 X 12.5 X 7 in.
Case of 4 Each for $277.03
Only null left
035210
View Documents Promotion Details
Sonoco™
613
15.5 X 15.5 X 12 in. 12.5 X 12.5 X 9 in.
Pack of 4 Each for $272.52
Only null left
03532101
View Documents Promotion Details
Sonoco™
TS615
15.5 X 15.5 X 15 in. 12.5 X 12.5 X 12 in.
Case of 3 Each for $228.24
Only null left
03001232
View Documents Promotion Details
Sonoco™
656
19.5 X 16.5 X 15.13 in. 16.75 X 13.75 X 12.63 in.
Pack of 2 Each for $250.21
Only null left
03001237
View Documents Promotion Details
Sonoco™
658
18.13 X 15.13 X 10.88 in. 15.81 X 12.81 X 8.5 in.
Pack of 4 Each for $476.00
Only null left
03001109
View Documents Promotion Details
Sonoco™
662
14.25 X 14.25 X 11.5 in. 12 X 12 X 9.75 in.
Case of 4 Each for $292.58
Only null left
Description
Description
  • Sizes that range 11 inches-17 inches in length
  • Also available in ThermoSafe Insulated EPS with Corrugated Containers and Coolers- Small (6  inches-10.5 inches in length)
  • Also available in ThermoSafe Insulated EPS with Corrugated Containers and Coolers- Large (19 inches-24 inches in length)
  • 200 lb. test corrugated
  • Excellent dry ice sublimation… average dry ice usage in 1-1/2 inch thick wall units is 5 pounds over 24 hours and 3-4 pounds thereafter.
  • Standard 1-1/2 inch wall shippers are ideal for refrigerated shipments for 24-48 hours.