Learn More
MilliporeSigma™ Supelco™ Thermogreen™ LB-2 Septa, Solid Discs
Suitable for conveniently introducing the sample without causing the system to leak. Improper septa decompose under the high-temperature conditions of capillary inlets causing baseline disruption and ghost peaks in a chromatogram.
Supplier:  MilliporeSigma™ Supelco™ 23157
 View more versions of this product
Catalog No. 111001601
Only null left
Description
Description
  • An improved version over the original Thermogreen™ LB-1 Septa
  • Extremely low bleed over a wide range of inlet temperatures (100° to 350°C)
  • Easy needle penetration
  • High puncture tolerance (ideal for autosamplers)
  • Rubber formulation exclusive to Supelco™
  • Pre-conditioned, ready to use
Specifications
Chemical Identifiers
Green
Solid Disc
Thermogreen
Rubber
100°C to 350°C
Septa, LB-2
Specifications
13/32 in.
10 mm
1000/Pk.
100°C to 350°C
Agilent/HP 5700 Series, Varian Packed Column Injectors
Green
Rubber
Solid Disc
Thermogreen
Septa, LB-2
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Thermogreen is a registered trademark of Sigma-Aldrich Co. LLC