Learn More
MilliporeSigma™ Supelco™ Thermogreen™ LB-2 Septa, Pre-drilled
For use with SPME applications to reduce septum coring
$287.78
Specifications
| For Use With (Application) | SPME |
|---|---|
| Style | Pre-drilled |
| Product Line | Thermogreen |
| Product Type | Septa, LB-2 |
| Catalog Number | Mfr. No. | Diameter (Metric) Outer | No. per Pack | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Diameter (Metric) Outer | No. per Pack | Price | Quantity & Availability | ||||
|
111020828
|
MilliporeSigma™ Supelco™
23168 |
11 mm | 50/Pk. |
Each for $287.78
|
|
||||
Description
For use with:
- Minimizing extraction onto or loss of analytes off the solid phase microextraction (SPME) fiber from the vial during analysis by GC-MS
- Avoiding carrier gas leaks, extraneous peaks, and phthalate contamination in GC instruments
- Use in conjunction with a thick needle protector to avoid damage to GC instruments irreversibly caused by microsyringes
- Influent sampling ports in gas chromatographs
Specifications
| SPME | |
| Pre-drilled | |
| Thermogreen | |
| Septa, LB-2 |
Thermogreen is a registered trademark of Sigma-Aldrich Co. LLC